Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291661.1 | NP_001278590.1 | 3 | 424 | ||
NM_198564.3 | NP_940966.2 | 3 | 424 | |||
XM_011533466.1 | XP_011531768.1 | 3 | 421 | |||
XM_011533467.1 | XP_011531769.1 | 3 | 421 | |||
XM_011533468.1 | XP_011531770.1 | 3 | 421 | |||
XM_011533469.1 | XP_011531771.1 | 3 | 421 | |||
XM_011533471.2 | XP_011531773.1 | 3 | 421 | |||
XM_011533474.1 | XP_011531776.1 | 3 | 421 | |||
XM_017005860.1 | XP_016861349.1 | 3 | 421 | |||
XM_017005861.1 | XP_016861350.1 | 3 | 1283 | |||
XM_017005863.1 | XP_016861352.1 | 3 | 421 | |||
GenBank mRNA | AK126276.1 | - | 3 | 421 |