Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: REEP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005669.4 | NP_005660.4 | 5 | 771 | ||
GenBank mRNA | AK294496.1 | - | 4 | 757 | ||
AK295436.1 | - | 4 | 523 | |||
AK298268.1 | - | 4 | 506 | |||
AK299902.1 | - | 6 | 1035 | |||
AK300048.1 | - | 5 | 589 | |||
AK310729.1 | - | 5 | 1135 | |||
BC000232.1 | - | 5 | 652 | 3350749 | ||
BC065926.1 | - | 5 | 630 | 6164621 | ||
BX648005.1 | - | 5 | 752 | |||
M74090.1 | - | 5 | 653 |