Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318060.1 | NP_001304989.1 | 11 | 1584 | ||
NM_001318061.1 | NP_001304990.1 | 12 | 1599 | |||
NM_001318062.1 | NP_001304991.1 | 12 | 1630 | |||
NM_006716.3 | NP_006707.1 | 12 | 1691 | |||
GenBank mRNA | AB028069.1 | - | 12 | 1704 | ||
AB028070.1 | - | 12 | 1643 | |||
AF160249.1 | - | 12 | 1403 | |||
AF160876.1 | - | 12 | 1386 | |||
AK022969.1 | - | 1 | 51 | |||
AK292569.1 | - | 12 | 1433 | |||
AK302010.1 | - | 11 | 1213 | |||
AK303175.1 | - | 12 | 1295 | |||
BC036045.1 | - | 12 | 1497 | 5273059 | ||
BC047693.1 | - | 12 | 1372 | 5753567 |