Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317769.1 | NP_001304698.1 | 7 | 796 | ||
NM_012108.3 | NP_036240.1 | 7 | 796 | |||
GenBank mRNA | AB023483.1 | - | 7 | 766 | ||
AK313676.1 | - | 7 | 745 | |||
BC014958.1 | - | 7 | 715 | 4851485 |