Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195555.1 | NP_001182484.1 | 6 | 750 | ||
NM_001195556.1 | NP_001182485.1 | 6 | 727 | |||
NM_014666.3 | NP_055481.1 | 6 | 750 | |||
XM_011534701.2 | XP_011533003.1 | 6 | 735 | |||
XM_017010087.1 | XP_016865576.1 | 6 | 735 | |||
XM_017010088.1 | XP_016865577.1 | 6 | 735 | |||
GenBank mRNA | AF434813.1 | - | 6 | 545 | ||
AK092765.1 | - | 6 | 697 | |||
AK300257.1 | - | 6 | 718 | |||
AL529948.3 | - | 6 | 737 | |||
BC004467.2 | - | 6 | 611 | 3831533 | ||
D79993.1 | - | 6 | 646 | |||
KU178486.1 | - | 6 | 545 | |||
KU178487.1 | - | 6 | 545 |