Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016323.3 | NP_057407.2 | 4 | 697 | ||
XM_011532022.2 | XP_011530324.2 | 4 | 1210 | |||
GenBank mRNA | AB027289.1 | - | 4 | 657 | ||
AY337518.1 | - | 4 | 697 | |||
BC140716.1 | - | 4 | 590 | 9021584 |