Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM206A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017832.3 | NP_060302.1 | 6 | 1041 | ||
GenBank mRNA | AF153417.1 | - | 6 | 971 | ||
AK000464.1 | - | 6 | 1025 | |||
AK310297.1 | - | 5 | 867 | |||
BC015795.1 | - | 6 | 1253 | 3910513 |