Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018356.2 | NP_060826.2 | 2 | 303 | ||
NR_134298.1 | 2 | 303 | ||||
XM_011514062.2 | XP_011512364.1 | 2 | 288 | |||
XM_017009607.1 | XP_016865096.1 | 2 | 292 | |||
XM_017009608.1 | XP_016865097.1 | 2 | 303 | |||
XM_017009610.1 | XP_016865099.1 | 2 | 292 | |||
XM_017009611.1 | XP_016865100.1 | 2 | 292 | |||
XM_017009613.1 | XP_016865102.1 | 2 | 483 | |||
XM_017009614.1 | XP_016865103.1 | 2 | 285 | |||
GenBank mRNA | AK002055.1 | - | 2 | 290 | ||
AK299156.1 | - | 2 | 280 | |||
BC021215.2 | - | 2 | 240 | 4302543 | ||
BC032845.1 | - | 2 | 286 | |||
DC404356.1 | - | 2 | 303 |