Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018359.3 | NP_060829.2 | 2, 3 | 191 | ||
NR_028085.1 | 2, 3 | 191 | ||||
GenBank mRNA | AK002062.1 | - | 2, 3 | 120 | ||
AK091635.1 | - | 2, 3 | 184 | |||
BC010493.1 | - | 1, 2 | 87 | 3048997 | ||
BP264462.1 | - | 2, 3 | 187 | |||
CR457278.1 | - | 1, 2 | 74 | |||
DA537391.1 | - | 2, 3 | 185 |