Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018409.3 | NP_060879.2 | 3 | 1087 | ||
XM_005263125.4 | XP_005263182.1 | 2 | 367 | |||
XM_006714260.3 | XP_006714323.1 | 4 | 1417 | |||
XM_006714265.3 | XP_006714328.1 | 3 | 477 | |||
XM_011532110.2 | XP_011530412.1 | 3 | 1290 | |||
XM_011532111.2 | XP_011530413.1 | 3 | 340 | |||
XM_017008410.1 | XP_016863899.1 | 4 | 2300 | |||
XM_017008411.1 | XP_016863900.1 | 3 | 908 | |||
GenBank mRNA | AB037746.1 | - | 3 | 1087 | ||
AK126913.1 | - | 4 | 1418 | |||
AL161975.1 | - | 2 | 2181 | |||
BC150161.1 | - | 3 | 594 | 40126591 | ||
BC152444.1 | - | 3 | 1087 | 8862572 |