Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG13 SIRNA Assays ›
Gene Symbol:
ALG13
Gene Name:
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases:
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location:
Chr.X: 111680741 - 111764164 on Build GRCh38
UniGene ID:
Hs.443061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
127166
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099922.2
NP_001093392.1
2
329
NM_001168385.2
NP_001161857.1
2
329
NM_001257230.1
NP_001244159.1
2
225
NM_001257234.1
NP_001244163.1
2
298
NM_001257235.2
NP_001244164.1
2
298
NM_001257237.1
NP_001244166.1
2
402
NM_001257239.2
NP_001244168.1
2
317
NM_001257240.2
NP_001244169.1
2
402
NM_001324290.1
NP_001311219.1
2
329
NM_001324291.1
NP_001311220.1
2
225
NM_001324292.1
NP_001311221.1
2
329
NM_001324293.1
NP_001311222.1
2
402
NM_001324294.1
NP_001311223.1
2
213
NM_018466.5
NP_060936.1
2
329
NR_033125.2
2
225
XM_006724693.2
XP_006724756.1
2
702
XM_006724697.3
XP_006724760.2
2
602
XM_006724698.3
XP_006724761.2
2
727
XM_011531028.2
XP_011529330.1
2
703
XM_011531030.1
XP_011529332.1
2
701
XM_011531031.1
XP_011529333.1
2
402
XM_011531032.1
XP_011529334.1
2
208
XM_011531033.2
XP_011529335.1
2
248
XM_017029846.1
XP_016885335.1
2
704
XM_017029847.1
XP_016885336.1
2
703
XM_017029848.1
XP_016885337.1
2
701
XM_017029849.1
XP_016885338.1
2
702
XM_017029850.1
XP_016885339.1
2
701
XM_017029851.1
XP_016885340.1
2
704
GenBank mRNA
AF220051.1
-
2
260
AK300394.1
-
2
167
AK302729.1
-
2
270
AK304712.1
-
2
219
AK312229.1
-
2
268
AK316306.1
-
2
271
AK316522.1
-
2
346
AL524204.3
-
2
148
BC005336.1
-
2
262
3996922
BE747995.1
-
2
258
BF240663.1
-
2
116
BF699152.1
-
2
330
BG621042.1
-
1
112
BM759764.1
-
2
311
CB268871.1
-
2
276
DA830058.1
-
2
393
DB292609.1
-
2
402
DC400909.1
-
2
213
DN992019.1
-
2
397
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
transferase
Gene Ontology Categories:
Function(s)
Process(es)
dolichol-linked oligosaccharide biosynthetic process
proteolysis
N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
thiol-dependent ubiquitinyl hydrolase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-8tvtn:80/10.239.165.222:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0