Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318163.1 | NP_001305092.1 | 16 | 1941 | ||
NM_001318164.1 | NP_001305093.1 | 16 | 2130 | |||
NM_019042.4 | NP_061915.2 | 16 | 2154 | |||
XM_005250462.3 | XP_005250519.1 | 17 | 2180 | |||
XM_011516336.2 | XP_011514638.1 | 16 | 2073 | |||
XM_017012367.1 | XP_016867856.1 | 17 | 2108 | |||
XM_017012368.1 | XP_016867857.1 | 14 | 1919 | |||
GenBank mRNA | AB067484.1 | - | 16 | 1868 | ||
AK000492.1 | - | 6 | 658 | |||
AK091283.1 | - | 16 | 2134 | |||
AK128629.1 | - | 16 | 2116 | |||
BC005209.2 | - | 8 | 867 | |||
BC011396.1 | - | 16 | 2110 | 3870711 |