Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020932.2 | NP_065983.1 | 1 | 1344 | ||
GenBank mRNA | AB046807.1 | - | 1 | 1273 | ||
AF490507.1 | - | 1 | 1273 | |||
AK291270.1 | - | 1 | 1259 | |||
BC050588.1 | - | 1 | 1225 | 6189864 |