Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022144.2 | NP_071427.2 | 7 | 1078 | ||
GenBank mRNA | AB055421.1 | - | 7 | 858 | ||
AF191770.1 | - | 7 | 1038 | |||
AF234259.1 | - | 7 | 919 | |||
AF291656.1 | - | 7 | 924 | |||
AY358706.1 | - | 7 | 910 |