Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C8orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023080.2 | NP_075568.1 | 5 | 1643 | ||
GenBank mRNA | AK024642.1 | - | 5 | 1641 | ||
BC010001.2 | - | 5 | 1603 | 4135341 | ||
BX648996.1 | - | 4 | 1703 |