Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRAMD3 SIRNA Assays ›
Gene Symbol:
GRAMD3
Gene Name:
GRAM domain containing 3
Gene Aliases:
NS3TP2
Chromosome Location:
Chr.5: 126360096 - 126494377 on Build GRCh38
UniGene ID:
Hs.363558
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
127352
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRAMD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146319.1
NP_001139791.1
4
651
NM_001146320.1
NP_001139792.1
3
688
NM_001146321.1
NP_001139793.1
5
843
NM_001146322.1
NP_001139794.1
4
462
NM_023927.2
NP_076416.2
4
800
XM_005272057.3
XP_005272114.1
4
538
XM_005272058.3
XP_005272115.1
4
538
XM_005272059.4
XP_005272116.1
5
928
XM_005272060.3
XP_005272117.1
4
625
XM_005272061.3
XP_005272118.1
4
620
XM_005272062.3
XP_005272119.1
4
586
XM_005272063.3
XP_005272120.1
4
735
XM_005272064.3
XP_005272121.1
5
645
XM_011543591.2
XP_011541893.1
5
1516
XM_011543592.2
XP_011541894.1
5
1512
XM_011543593.2
XP_011541895.1
4
625
XM_011543594.2
XP_011541896.1
4
397
XM_011543595.2
XP_011541897.1
3
392
XM_017009781.1
XP_016865270.1
5
403
XM_017009782.1
XP_016865271.1
5
645
XM_017009783.1
XP_016865272.1
3
256
XM_017009784.1
XP_016865273.1
3
444
XM_017009785.1
XP_016865274.1
3
497
GenBank mRNA
AK024966.1
-
4
568
AK225829.1
-
4
568
AK293307.1
-
3
393
AK294656.1
-
4
450
AK296267.1
-
5
548
AK298009.1
-
4
385
AK298349.1
-
4
473
AK300154.1
-
4
596
AK303857.1
-
4
402
AK316288.1
-
4
615
AY116970.1
-
4
370
BC008590.1
-
4
533
4148772
DC413835.1
-
4
416
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0