Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272059.4 | XP_005272116.1 | 15 | 3107 | ||
XM_005272060.3 | XP_005272117.1 | 14 | 2804 | |||
XM_005272061.3 | XP_005272118.1 | 14 | 2799 | |||
XM_005272063.3 | XP_005272120.1 | 14 | 2911 | |||
XM_005272064.3 | XP_005272121.1 | 15 | 2824 | |||
XM_011543591.2 | XP_011541893.1 | 15 | 3695 | |||
XM_011543592.2 | XP_011541894.1 | 15 | 3688 | |||
XM_011543593.2 | XP_011541895.1 | 14 | 2804 | |||
XM_011543594.2 | XP_011541896.1 | 14 | 2576 | |||
XM_011543595.2 | XP_011541897.1 | 13 | 2571 | |||
XM_017009781.1 | XP_016865270.1 | 15 | 2582 | |||
XM_017009782.1 | XP_016865271.1 | 15 | 2821 | |||
XM_017009784.1 | XP_016865273.1 | 13 | 2623 | |||
XM_017009785.1 | XP_016865274.1 | 13 | 2673 | |||
GenBank mRNA | AK024966.1 | - | 14 | 2744 | ||
AL157454.1 | - | 1 | 867 |