Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257998.1 | NP_001244927.1 | 12 | 929 | ||
NM_017667.3 | NP_060137.2 | 11 | 901 | |||
NM_024553.2 | NP_078829.1 | 11 | 901 | |||
XM_011516395.2 | XP_011514697.1 | 11 | 891 | |||
XM_011516396.2 | XP_011514698.1 | 11 | 898 | |||
GenBank mRNA | AB058764.2 | - | 11 | 847 | ||
AB100163.1 | - | 11 | 858 | |||
AK000104.1 | - | 3 | 133 | |||
AK055965.1 | - | 11 | 865 | |||
AK126478.1 | - | 12 | 903 | |||
AL832393.1 | - | 4 | 481 | |||
AL833112.1 | - | 10 | 769 | |||
BC017888.1 | - | 11 | 901 | |||
BC047757.1 | - | 3 | 332 | |||
BC108708.1 | - | 11 | 863 | |||
BC132740.1 | - | 11 | 847 | |||
BC132742.1 | - | 11 | 847 | |||
BC152424.1 | - | 11 | 847 | 8862538 |