Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024590.3 | NP_078866.3 | 2 | 4276 | ||
XM_005263212.4 | XP_005263269.1 | 3 | 4138 | |||
XM_011532238.2 | XP_011530540.1 | 3 | 3613 | |||
XM_011532239.2 | XP_011530541.1 | 4 | 3151 | |||
XM_017008592.1 | XP_016864081.1 | 3 | 4316 | |||
XM_017008593.1 | XP_016864082.1 | 6 | 4871 | |||
XM_017008594.1 | XP_016864083.1 | 5 | 4438 | |||
XM_017008595.1 | XP_016864084.1 | 6 | 4551 | |||
XM_017008596.1 | XP_016864085.1 | 7 | 4435 | |||
XM_017008597.1 | XP_016864086.1 | 6 | 4562 | |||
XM_017008598.1 | XP_016864087.1 | 4 | 4388 | |||
XM_017008599.1 | XP_016864088.1 | 4 | 4397 | |||
XM_017008600.1 | XP_016864089.1 | 3 | 3101 | |||
GenBank mRNA | AY358647.1 | - | 4 | 4137 |