Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170796.1 | NP_001164267.1 | 8 | 721 | ||
NM_024699.2 | NP_078975.2 | 8 | 742 | |||
NR_033193.1 | 8 | 744 | ||||
NR_033194.1 | 7 | 702 | ||||
NR_033195.1 | 7 | 680 | ||||
GenBank mRNA | AK024069.1 | - | 8 | 733 | ||
AK225195.1 | - | 8 | 717 | |||
AK304111.1 | - | 7 | 685 | |||
AL833030.1 | - | 4 | 1270 | |||
BC022251.2 | - | 8 | 727 | 4695118 | ||
BC051857.1 | - | 7 | 684 | 6012307 | ||
BC056877.1 | - | 7 | 685 | 6140929 | ||
BQ427843.1 | - | 8 | 742 | |||
CR457323.1 | - | 8 | 719 |