Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170796.1 | NP_001164267.1 | 8 | 1626 | ||
NM_024699.2 | NP_078975.2 | 8 | 1647 | |||
NR_033193.1 | 8 | 1649 | ||||
NR_033194.1 | 7 | 1607 | ||||
NR_033195.1 | 7 | 1585 | ||||
GenBank mRNA | AK024069.1 | - | 8 | 1638 | ||
AK225195.1 | - | 8 | 1622 | |||
AL833030.1 | - | 4 | 2175 | |||
BC051857.1 | - | 7 | 1589 | 6012307 | ||
BC056877.1 | - | 7 | 1589 | 6140929 |