Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPCAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024830.3 | NP_079106.3 | 14 | 3310 | ||
XM_005248373.3 | XP_005248430.1 | 15 | 3218 | |||
XM_011514132.1 | XP_011512434.1 | 14 | 3319 | |||
XM_011514134.1 | XP_011512436.1 | 14 | 3085 | |||
XM_017009879.1 | XP_016865368.1 | 8 | 2621 | |||
GenBank mRNA | AF052162.1 | - | 1 | 983 | ||
AK022499.1 | - | 9 | 2488 | |||
AK022505.1 | - | 9 | 2488 | |||
AK090444.1 | - | 12 | 2789 | |||
AK095552.1 | - | 3 | 2061 | |||
AK123603.1 | - | 1 | 3424 | |||
AL831864.1 | - | 2 | 1833 | |||
BC002543.2 | - | 1 | 686 | |||
BC020166.1 | - | 12 | 2776 |