Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGIX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099681.2 | NP_001093151.2 | 5 | 1298 | ||
NM_001099682.2 | NP_001093152.2 | 5 | 1283 | |||
NM_024859.3 | NP_079135.3 | 6 | 1526 | |||
GenBank mRNA | AK025340.1 | - | 4 | 1381 |