Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CLPTM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030782.4 | NP_110409.2 | 17 | 1876 | ||
XM_011514144.2 | XP_011512446.1 | 17 | 1840 | |||
GenBank mRNA | AB045223.1 | - | 17 | 1633 | ||
AK027306.1 | - | 17 | 1690 | |||
AK057095.1 | - | 1 | 1112 | |||
AK126225.1 | - | 1 | 2994 | |||
AK128462.1 | - | 8 | 3069 | |||
AL137440.1 | - | 1 | 211 | |||
AL832953.1 | - | 7 | 478 | |||
BC016399.1 | - | 14 | 1154 | |||
BC025305.1 | - | 17 | 1694 | 3051368 | ||
BC040705.1 | - | 1 | 1002 |