Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033035.4 | NP_149024.1 | 3 | 449 | ||
NM_138551.4 | NP_612561.2 | 1 | 231 | |||
NR_045089.1 | 4 | 1654 | ||||
XM_011543698.1 | XP_011542000.1 | 2 | 53 | |||
GenBank mRNA | AF338732.1 | - | 3 | 283 | ||
AL833421.1 | - | 1 | 211 | |||
AY037115.1 | - | 3 | 449 | |||
BC016720.1 | - | 1 | 164 | |||
BC022894.1 | - | 1 | 203 | |||
BC040592.1 | - | 4 | 1655 | 5269764 | ||
BF245025.1 | - | 1 | 218 | |||
BU571002.1 | - | 1 | 234 |