Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSKH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033126.1 | NP_149117.1 | 2 | 715 | ||
XM_017013929.1 | XP_016869418.1 | 4 | 1072 | |||
GenBank mRNA | AY037806.1 | - | 2 | 715 | ||
BC126180.1 | - | 2 | 790 | 8991896 | ||
BC126182.1 | - | 2 | 790 | 8991898 |