Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195639.1 | NP_001182568.1 | 4 | 943 | ||
NM_052958.2 | NP_443190.2 | 4 | 943 | |||
XM_011517445.2 | XP_011515747.1 | 4 | 915 | |||
XM_011517446.2 | XP_011515748.1 | 4 | 915 | |||
XM_011517449.2 | XP_011515751.1 | 4 | 915 | |||
XM_011517450.2 | XP_011515752.1 | 4 | 915 | |||
GenBank mRNA | AB056652.1 | - | 3 | 1911 | ||
AK291436.1 | - | 4 | 415 | |||
AL834454.1 | - | 4 | 440 | |||
BC041961.1 | - | 4 | 943 | 5302156 |