Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199380.1 | NP_001186309.1 | 3, 4 | 541 | ||
NM_001199381.1 | NP_001186310.1 | 3, 4 | 588 | |||
NM_001199382.1 | NP_001186311.1 | 3, 4 | 514 | |||
NM_001199383.1 | NP_001186312.1 | 3, 4 | 665 | |||
NM_144726.2 | NP_653327.1 | 3, 4 | 412 | |||
XM_017009138.1 | XP_016864627.1 | 3, 4 | 860 | |||
GenBank mRNA | AK056513.1 | - | 3, 4 | 412 | ||
AK292854.1 | - | 3, 4 | 654 | |||
AK304228.1 | - | 3, 4 | 500 | |||
AK304435.1 | - | 3, 4 | 588 | |||
AK308394.1 | - | 3, 4 | 541 | |||
AK311433.1 | - | 3, 4 | 526 | |||
BC042684.1 | - | 3, 4 | 677 | 4826810 | ||
BC044767.1 | - | 3, 4 | 677 | |||
DC418555.1 | - | 3, 4 | 515 |