Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDUFAF6 SIRNA Assays ›
Gene Symbol:
NDUFAF6
Gene Name:
NADH:ubiquinone oxidoreductase complex assembly factor 6
Gene Aliases:
C8orf38
Chromosome Location:
Chr.8: 94895271 - 95118496 on Build GRCh38
UniGene ID:
Hs.729144
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
127981
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDUFAF6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152416.3
NP_689629.2
6
619
XM_005250789.2
XP_005250846.1
6
630
XM_005250790.2
XP_005250847.1
7
730
XM_005250791.2
XP_005250848.1
7
696
XM_005250792.2
XP_005250849.1
7
700
XM_005250793.2
XP_005250850.1
7
613
XM_011516833.2
XP_011515135.1
11
1839
XM_011516834.2
XP_011515136.1
10
1800
XM_011516835.2
XP_011515137.1
10
1058
XM_011516836.2
XP_011515138.1
10
1741
XM_011516837.2
XP_011515139.1
11
1799
XM_011516838.2
XP_011515140.1
10
1676
XM_011516839.2
XP_011515141.1
9
1578
XM_011516840.2
XP_011515142.1
10
1672
XM_011516841.2
XP_011515143.1
10
1701
XM_011516842.2
XP_011515144.1
9
1574
XM_017013027.1
XP_016868516.1
10
1050
XM_017013028.1
XP_016868517.1
9
950
XM_017013029.1
XP_016868518.1
8
1539
XM_017013030.1
XP_016868519.1
8
1284
XM_017013031.1
XP_016868520.1
9
1108
XM_017013032.1
XP_016868521.1
7
730
XM_017013033.1
XP_016868522.1
6
427
XM_017013034.1
XP_016868523.1
5
400
XM_017013035.1
XP_016868524.1
9
1152
XM_017013036.1
XP_016868525.1
9
1484
XM_017013037.1
XP_016868526.1
7
610
XM_017013038.1
XP_016868527.1
8
679
XM_017013039.1
XP_016868528.1
7
836
GenBank mRNA
AA625387.1
-
2
83
AK298631.1
-
6
539
AK301414.1
-
7
661
AY444560.1
-
7
690
BC028166.1
-
8
952
BF030925.1
-
2
131
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001085493, AK009547.
Gene Ontology Categories:
Function(s)
Process(es)
biosynthetic process
mitochondrial respiratory chain complex I assembly
transmembrane transport
transferase activity
transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0