- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: TMEM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286657.1 | NP_001273586.1 | 2, 3 | 435 | ||
NM_152417.2 | NP_689630.1 | 3, 4 | 574 | |||
XM_005251150.1 | XP_005251207.1 | 3, 4 | 563 | |||
XM_011517454.1 | XP_011515756.1 | 3, 4 | 587 | |||
XM_011517455.2 | XP_011515757.1 | 3, 4 | 563 | |||
GenBank mRNA | AK056932.1 | - | 3, 4 | 493 | ||
AL832935.1 | - | 1, 2 | 44 |