Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003145.3 | NP_003136.1 | 4 | 385 | ||
XM_011509923.2 | XP_011508225.1 | 4 | 410 | |||
GenBank mRNA | AK222600.1 | - | 4 | 328 | ||
AK300678.1 | - | 4 | 385 | |||
AK313821.1 | - | 4 | 385 | |||
AY251536.1 | - | 4 | 345 | |||
BC000341.2 | - | 4 | 331 | 2822983 | ||
BP196192.1 | - | 4 | 385 | |||
BX649192.1 | - | 4 | 370 | |||
CR456973.1 | - | 3 | 305 | |||
D37991.1 | - | 4 | 329 | |||
X74104.1 | - | 4 | 355 |