Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169574.1 | NP_001163045.1 | 3, 4 | 566 | ||
NM_152761.2 | NP_689974.2 | 3, 4 | 566 | |||
XM_011545473.2 | XP_011543775.1 | 3, 4 | 323 | |||
GenBank mRNA | AK058173.1 | - | 3, 4 | 566 | ||
BC117344.1 | - | 3, 4 | 478 | 40125895 | ||
BC117346.1 | - | 3, 4 | 478 | 40125897 | ||
BC144007.1 | - | 3, 4 | 478 | 9052526 |