Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 19 | 2731 | ||
NM_153223.3 | NP_694955.2 | 20 | 2812 | |||
XM_005271901.4 | XP_005271958.1 | 19 | 2593 | |||
XM_011543185.2 | XP_011541487.1 | 19 | 3146 | |||
XM_011543186.2 | XP_011541488.1 | 16 | 2425 | |||
XM_017009085.1 | XP_016864574.1 | 16 | 2290 | |||
GenBank mRNA | AK093409.1 | - | 12 | 1498 | ||
AK296999.1 | - | 18 | 2506 | |||
AL833929.1 | - | 8 | 920 | |||
BC040527.1 | - | 20 | 3589 | |||
BC144558.1 | - | 22 | 2939 | |||
BX648687.1 | - | 10 | 1205 |