Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145128.2 | NP_001138600.2 | 38, 39 | 5400 | ||
XM_006715376.3 | XP_006715439.2 | 39, 40 | 6265 | |||
XM_011535550.2 | XP_011533852.1 | 39, 40 | 6280 | |||
XM_011535552.2 | XP_011533854.1 | 38, 39 | 5456 | |||
XM_011535553.2 | XP_011533855.1 | 38, 39 | 6063 | |||
XM_011535554.2 | XP_011533856.1 | 37, 38 | 5959 | |||
XM_011535558.1 | XP_011533860.1 | 20, 21 | 3301 | |||
XM_017010385.1 | XP_016865874.1 | 38, 39 | 5448 | |||
XM_017010387.1 | XP_016865876.1 | 33, 34 | 5911 | |||
XM_017010389.1 | XP_016865878.1 | 24, 25 | 4163 | |||
GenBank mRNA | AK098657.1 | - | 4, 5 | 638 | ||
BC101714.1 | - | 4, 5 | 566 | 8069220 | ||
BC111948.1 | - | 4, 5 | 566 | 8327416 | ||
BC143533.1 | - | 4, 5 | 524 | 9052042 |