Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174952.2 | NP_777612.1 | 1, 2 | 200 | ||
XM_011531886.2 | XP_011530188.1 | 1, 2 | 433 | |||
XM_017008049.1 | XP_016863538.1 | 1, 2 | 433 | |||
XM_017008050.1 | XP_016863539.1 | 1, 2 | 433 | |||
GenBank mRNA | BC036870.1 | - | 1, 2 | 200 | 5226859 |