Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVOPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139456.1 | NP_001132928.1 | 11 | 1126 | ||
NM_174959.3 | NP_777619.1 | 7 | 743 | |||
XM_005250143.3 | XP_005250200.1 | 12 | 1530 | |||
XM_011515797.2 | XP_011514099.1 | 8 | 817 | |||
XM_017011746.1 | XP_016867235.1 | 9 | 932 | |||
XM_017011747.1 | XP_016867236.1 | 8 | 757 | |||
XM_017011748.1 | XP_016867237.1 | 8 | 762 | |||
GenBank mRNA | BC036796.1 | - | 7 | 743 | 5590416 | |
BX349158.2 | - | 2 | 173 | |||
BX648789.1 | - | 7 | 4363 |