Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181489.5 | NP_852466.1 | 3 | 536 | ||
XM_005265102.2 | XP_005265159.1 | 2 | 394 | |||
XM_011533674.2 | XP_011531976.1 | 2 | 359 | |||
XM_017006345.1 | XP_016861834.1 | 2 | 451 | |||
XM_017006346.1 | XP_016861835.1 | 2 | 394 | |||
GenBank mRNA | AK310588.1 | - | 2 | 395 | ||
AY262260.1 | - | 1 | 339 | |||
AY279350.1 | - | 2 | 355 | |||
AY295873.2 | - | 3 | 536 | |||
BC101486.1 | - | 1 | 250 | 8068992 | ||
BC143439.1 | - | 1 | 251 | 9051947 | ||
L32162.1 | - | 2 | 355 |