Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FN3KRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024619.3 | NP_078895.2 | 1, 2 | 199 | ||
GenBank mRNA | AK022233.1 | - | 1, 2 | 166 | ||
AK293523.1 | - | 1, 2 | 183 | |||
AK315373.1 | - | 1, 2 | 183 | |||
AL136631.1 | - | 1, 2 | 170 | |||
AY360465.1 | - | 1, 2 | 166 | |||
BC007611.1 | - | 1, 2 | 175 | 3351601 | ||
BC014408.1 | - | 1, 2 | 153 | 4652087 | ||
CR533461.1 | - | 1, 2 | 149 | |||
DA019669.1 | - | 1, 2 | 200 |