Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257967.1 | NP_001244896.1 | 8 | 675 | ||
NM_001257968.1 | NP_001244897.1 | 7 | 700 | |||
NM_194300.3 | NP_919276.2 | 6 | 563 | |||
NR_047565.1 | 7 | 1521 | ||||
XM_011515213.1 | XP_011513515.1 | 7 | 667 | |||
XM_017011871.1 | XP_016867360.1 | 7 | 660 | |||
XM_017011872.1 | XP_016867361.1 | 7 | 667 | |||
GenBank mRNA | AK095663.1 | - | 5 | 436 | ||
AK128026.1 | - | 7 | 1521 | |||
AK295011.1 | - | 7 | 618 | |||
AK304387.1 | - | 8 | 675 | |||
BC132719.1 | - | 7 | 566 | 40146741 |