Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291593.1 | NP_001278522.1 | 3, 4 | 555 | ||
NM_015102.4 | NP_055917.1 | 3, 4 | 555 | |||
NR_111987.1 | 3, 4 | 555 | ||||
XM_006710563.3 | XP_006710626.1 | 3, 4 | 401 | |||
XM_011541213.1 | XP_011539515.1 | 2, 3 | 325 | |||
XM_011541214.1 | XP_011539516.1 | 2, 3 | 325 | |||
XM_011541215.1 | XP_011539517.1 | 2, 3 | 325 | |||
XM_011541216.2 | XP_011539518.1 | 4, 5 | 1029 | |||
XM_011541217.2 | XP_011539519.1 | 3, 4 | 893 | |||
XM_011541218.2 | XP_011539520.1 | 4, 5 | 562 | |||
XM_017000996.1 | XP_016856485.1 | 2, 3 | 325 | |||
XM_017000997.1 | XP_016856486.1 | 3, 4 | 824 | |||
XM_017000998.1 | XP_016856487.1 | 2, 3 | 325 | |||
XM_017000999.1 | XP_016856488.1 | 3, 4 | 320 | |||
XM_017001002.1 | XP_016856491.1 | 2, 3 | 325 | |||
GenBank mRNA | AF537130.1 | - | 3, 4 | 553 | ||
AK314326.1 | - | 3, 4 | 529 | |||
AY118228.1 | - | 3, 4 | 490 | |||
BC040520.1 | - | 3, 4 | 556 | 5295140 |