Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NPHP4 SIRNA Assays ›
Gene Symbol:
NPHP4
Gene Name:
nephronophthisis 4
Gene Aliases:
POC10, SLSN4
Chromosome Location:
Chr.1: 5862808 - 5992473 on Build GRCh38
UniGene ID:
Hs.462348
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
128696
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NPHP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291593.1
NP_001278522.1
16, 17
2569
NM_001291594.1
NP_001278523.1
15, 16
2392
NM_015102.4
NP_055917.1
19, 20
2879
XM_006710563.3
XP_006710626.1
19, 20
2725
XM_011541213.1
XP_011539515.1
18, 19
2646
XM_011541214.1
XP_011539516.1
18, 19
2649
XM_011541215.1
XP_011539517.1
17, 18
2538
XM_011541216.2
XP_011539518.1
20, 21
3353
XM_011541217.2
XP_011539519.1
19, 20
3217
XM_011541218.2
XP_011539520.1
20, 21
2886
XM_017000996.1
XP_016856485.1
18, 19
2646
XM_017000997.1
XP_016856486.1
19, 20
3148
XM_017000998.1
XP_016856487.1
18, 19
2649
XM_017000999.1
XP_016856488.1
19, 20
2644
XM_017001000.1
XP_016856489.1
18, 19
2477
XM_017001001.1
XP_016856490.1
13, 14
1916
XM_017001002.1
XP_016856491.1
18, 19
2649
XM_017001003.1
XP_016856492.1
11, 12
1707
GenBank mRNA
AB014573.1
-
14, 15
1979
AF537130.1
-
19, 20
2877
AK124074.1
-
15, 16
2345
AK128182.1
-
1, 2
2736
AK314326.1
-
16, 17
2543
AY118228.1
-
19, 20
2814
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
visual behavior
single organismal cell-cell adhesion
actin cytoskeleton organization
sperm motility
hippo signaling
photoreceptor cell outer segment organization
photoreceptor cell maintenance
retina development in camera-type eye
negative regulation of canonical Wnt signaling pathway
positive regulation of bicellular tight junction assembly
structural molecule activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0