Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287011.1 | NP_001273940.1 | 4 | 485 | ||
NM_001287013.1 | NP_001273942.1 | 4 | 322 | |||
NM_022343.3 | NP_071738.1 | 4 | 485 | |||
NR_104637.1 | 6 | 866 | ||||
NR_104638.1 | 4 | 277 | ||||
NR_104639.1 | 4 | 485 | ||||
NR_104640.1 | 4 | 507 | ||||
GenBank mRNA | AJ011129.1 | - | 3 | 242 | ||
AK074488.1 | - | 4 | 307 | |||
AK075091.1 | - | 4 | 286 | |||
AK091288.1 | - | 5 | 1099 | |||
AK298741.1 | - | 4 | 286 | |||
AK307243.1 | - | 4 | 286 | |||
AY039756.1 | - | 4 | 276 | |||
BC017918.1 | - | 4 | 288 | 4296721 | ||
CR749781.1 | - | 6 | 667 | |||
DA268281.1 | - | 4 | 322 | |||
DA271525.1 | - | 4 | 262 | |||
DA824243.1 | - | 4 | 276 | |||
DA986727.1 | - | 4 | 286 |