Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030630.2 | NP_085133.1 | 2 | 237 | ||
XM_005257226.1 | XP_005257283.1 | 2 | 235 | |||
GenBank mRNA | AK074401.1 | - | 2 | 166 | ||
AK095565.1 | - | 2 | 195 | |||
AK304081.1 | - | 2 | 152 | |||
AL137556.1 | - | 2 | 184 | |||
BC035372.1 | - | 2 | 153 | 5174235 | ||
DA336027.1 | - | 2 | 237 |