Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF32 SIRNA Assays ›
Gene Symbol:
RNF32
Gene Name:
ring finger protein 32
Gene Aliases:
FKSG33, HSD15, LMBR2
Chromosome Location:
Chr.7: 156640286 - 156678333 on Build GRCh38
UniGene ID:
Hs.446194
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
128780
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF32
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184996.1
NP_001171925.1
6
867
NM_001184997.1
NP_001171926.1
6
720
NM_001308273.1
NP_001295202.1
6
818
NM_001308274.1
NP_001295203.1
5
609
NM_030936.3
NP_112198.1
6
762
NR_131778.1
6
716
XM_005249522.4
XP_005249579.1
6
674
XM_011515804.2
XP_011514106.1
7
784
XM_011515805.2
XP_011514107.1
7
941
XM_011515806.2
XP_011514108.1
6
852
XM_011515807.2
XP_011514109.1
6
660
XM_011515808.2
XP_011514110.1
6
772
XM_011515809.2
XP_011514111.1
6
845
XM_011515810.2
XP_011514112.1
6
662
XM_011515811.2
XP_011514113.1
6
657
XM_011515812.2
XP_011514114.1
6
764
XM_011515813.2
XP_011514115.1
6
723
XM_017011753.1
XP_016867242.1
6
766
XM_017011754.1
XP_016867243.1
6
766
XM_017011755.1
XP_016867244.1
6
766
GenBank mRNA
AF325690.1
-
6
670
AF441222.1
-
5
609
AF441224.1
-
6
716
AK307316.1
-
7
834
AK314067.1
-
6
867
AL136874.1
-
6
650
BC015416.1
-
5
620
4395421
BT007037.1
-
4
532
CR533513.1
-
4
532
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-dvnl5:80/10.239.162.83:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0