Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033515.2 | NP_277050.2 | 2 | 271 | ||
XM_005267212.2 | XP_005267269.2 | 2 | 347 | |||
XM_005267213.1 | XP_005267270.1 | 2 | 271 | |||
XM_017011494.1 | XP_016866983.1 | 2 | 347 | |||
GenBank mRNA | AB053293.1 | - | 1 | 69 | ||
AK098594.1 | - | 2 | 271 | |||
AK307796.1 | - | 4 | 346 | |||
AL834511.1 | - | 2 | 209 | |||
AY643712.1 | - | 2 | 272 | |||
BC017223.2 | - | 2 | 193 | |||
BC039611.1 | - | 2 | 200 | |||
BC062417.1 | - | 2 | 180 | |||
BC101708.1 | - | 2 | 265 | 8069214 | ||
BC107416.1 | - | 2 | 227 | 4040931 | ||
BC111940.1 | - | 2 | 265 | 8327408 | ||
BC143531.1 | - | 2 | 265 | |||
BM557223.1 | - | 2 | 208 | |||
CB149638.1 | - | 2 | 271 |