Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC18B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052831.2 | NP_439896.1 | 14 | 1502 | ||
XM_017010236.1 | XP_016865725.1 | 14 | 1693 | |||
GenBank mRNA | AJ420537.1 | - | 8 | 648 | ||
AK124442.1 | - | 14 | 1381 | |||
AK289918.1 | - | 14 | 1521 | |||
BC069567.1 | - | 10 | 905 | |||
BC069570.1 | - | 14 | 1422 | 7262298 |