Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052876.3 | NP_443108.1 | 6 | 2981 | ||
XM_005259721.3 | XP_005259778.1 | 7 | 3040 | |||
GenBank mRNA | AK094702.1 | - | 2 | 1581 | ||
AK123056.1 | - | 1 | 279 | |||
AK226064.1 | - | 4 | 1788 | |||
BC055396.1 | - | 7 | 2913 | 6142288 |