Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052886.2 | NP_443118.1 | 3, 4 | 557 | ||
XM_011516807.2 | XP_011515109.1 | 2, 3 | 341 | |||
GenBank mRNA | AK172820.1 | - | 3, 4 | 473 | ||
AK312030.1 | - | 3, 4 | 464 | |||
AY007723.1 | - | 3, 4 | 534 | |||
BC012367.1 | - | 3, 4 | 557 | 3860009 | ||
CV574204.1 | - | 3, 4 | 557 |