Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CYYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320768.1 | NP_001307697.1 | 4 | 1240 | ||
NM_052954.3 | NP_443186.2 | 4 | 1237 | |||
NR_135472.1 | 5 | 1407 | ||||
GenBank mRNA | AF401639.1 | - | 4 | 842 | ||
AK054581.1 | - | 4 | 1117 | |||
AK223576.1 | - | 4 | 957 | |||
AK304124.1 | - | 5 | 1287 | |||
AL833200.1 | - | 4 | 1048 | |||
AY061853.1 | - | 4 | 782 | |||
BC036761.1 | - | 4 | 1025 | 5588436 |