Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080387.4 | NP_525126.2 | 6 | 893 | ||
XM_011520632.2 | XP_011518934.1 | 7 | 862 | |||
GenBank mRNA | AY115592.1 | - | 6 | 884 | ||
AY297446.1 | - | 6 | 891 | |||
BC032313.1 | - | 6 | 893 | 5218660 |